Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-AQP4 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

600-401-E92
ROCK600-401-E92EA 763 EUR InStock
ROCK600-401-E92
Anti-AQP4 Rabbit Polyclonal Antibody
Antikörper
Anti-Aquaporin 4 Antibody is suitable for use in WB and IF microscopy. Expect a band approximately ~35kDa on specific lysates. May detect larger glycosylated bands ~50kDa. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: Immunofluorescence: 1:400; Western Blot: 1:1000-4000

Type: Primary
Antigen: AQP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat, Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge