Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-mCherry Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

600-401-P16S
ROCK600-401-P16SEA 226 EUR InStock
ROCK600-401-P16S
Anti-mCherry Rabbit Polyclonal Antibody
Antikörper
mCherry antibody is ideal for western blotting. Fluorescent proteins such as Discosoma Red Fluorescent Protein (and its varients), and GFP are widely used in research practice. Both commonly serve as a markers for gene expression and protein localization. DsRed was isolated from sea anemone Discosoma sp. mushroom and GFP is originated from Aequorea victoria jellyfish. As DsRed and GFP share only 19% identity, therefore, in general, anti-GFP antibodies do not recognize DsRed protein and vice versa. Structurally, Discosoma red fluorescent protein is similar to Aequorea green fluorescent protein in terms of its overall fold (a β-can) and chromophore-formation chemistry. However, Discosoma red fluorescent protein undergoes an additional steps in the chromophore maturation and obligates tetrameric structure. All mCherry antibodies have been pre-absorbed to eliminate any potential cross-reactivity to human, mouse and rat serum proteins. The antibodies are also confirmed for non-reactivity to GFP protein.

contains 0.01% (w/v) Sodium Azide

Type: Tag
Antigen: mCherry
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge