Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-MeK Rabbit Polyclonal Antibody (Biotin)

Lieferant: Rockland Immunochemicals

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

600-406-940
ROCK600-406-940EA 861 EUR InStock
ROCK600-406-940
Anti-MeK Rabbit Polyclonal Antibody (Biotin)
Antikörper
This affinity-purified antibody is suitable for use in ELISA and western blotting assays. The antibody reacts specifically with methylated lysine residues. Although not tested, this antibody is likely functional in RIA, flow cytometry and immunohistochemistry.

Recommended Dilutions: ELISA: 1:5000; Western Blot: 1:1000;

Type: Primary
Antigen: MeK
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mammalian, Avian,

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge