Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-kappa light chain Goat Polyclonal Antibody

Lieferant: Rockland Immunochemicals

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

609-1110
ROCK609-1110EA 324 EUR InStock
ROCK609-1110
Anti-kappa light chain Goat Polyclonal Antibody
Antikörper
Human kappa chain Antibody is suitable for immunoassays where specificity to the immunoglobulin kappa subuint is desired. Antibody has been tested by ELISA, western blot immunoblot, and immunohistochemisty.

Recommended Dilutions: ELISA: 1:10000 - 1:20000; Western Blot: 1:1000 - 1:10000; Immunochemistry: 1:500 - 1:2000;

Type: Secondary
Antigen: Igκ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Light Chain
Host: Goat
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge