Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-IgG F(c) Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Rockland Immunochemicals

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

609-1303
ROCK609-1303EA 341 EUR InStock
ROCK609-1303
Anti-IgG F(c) Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Anti-HUMAN IgG F(c) (GOAT) Antibody has been assayed against 1.0 µg of Human IgG in a standard capture ELISA using ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid)) as a substrate for 30 minutes at room temperature.

Recommended Dilutions: ELISA: 1:60,000; Western Blot: 1:1000 - 1:5000; Immunochemistry: 1:500 - 1:2500;
Restore with deionized water (or equivalent); 1.0 ml
Do not add Sodium Azide!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: F(c)
Host: Goat
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge