NF[kappa]B transcription factor assay kits
Lieferant: ROCKLAND
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
KAA064
ROCKKAA064EA 1490 EUR InStock
ROCKKAA064
NF[kappa]B transcription factor assay kits
Tests Zelltests
These assays are non-radioactive methods for detecting specific transcription factor DNA binding activity in nuclear extracts and whole cell lysates. A specific dsDNA sequence containing the NF[kappa]B response element is immobilized on the bottom of the wells in a 96-well plate. The transcription factor is detected by the addition of a specific primary antibody. A colorimetric readout at 450 nm is obtained through a horseradish peroxidase-conjugated secondary antibody.
Lieferumfang: Includes transcription factor A, NF[kappa]B p50 or NF[kappa]B p65 positive control and primary antibody, HRP-conjugated goat anti-rabbit secondary antibody, specific competitor dsDNA, antibody binding buffer (10X), binding assay (4X), wash buffer concentrate (400X), Tween®-20 polysorbate, developing solution, stop solution, and 96-well plate and plate cover.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge