2X SDS-PAGE sample buffer without DTT or b-ME
Lieferant: ROCKLAND
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
MB-018
ROCKMB-018EA 170 EUR InStock
ROCKMB-018
2X SDS-PAGE sample buffer without DTT or b-ME
Puffer Elektrophorese-Puffer Beladungspuffer
Sample buffer for protein electrophoresis.
This product is a concentrated stock solution and should be diluted appropriately with distilled, deionised water or equivalent to its final working concentration. 2X SDS-PAGE sample buffer consists of 0,125 M Tris, 4% (w/v) SDS, 20% (v/v) glycerol and 0,01% (w/v) bromophenol blue. The pH of this solution is 6,8. Meticulously prepared using ultra pure reagents dissolved in highly polished pharmaceutical grade deionised water.
This product was aseptically filtered through a Millipore 0,22 µm filter into clean, pre-sterilised containers. The product was tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria.
Achtung! Bitte beachten: This product is for research use only and is not intended for therapeutic or diagnostic applications.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge