Position TrueBlot® magnetic separator, 6
Lieferant: Rockland Immunochemicals
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
TMS-15-50
ROCKTMS-15-50EA 847 EUR InStock
ROCKTMS-15-50
Position TrueBlot® magnetic separator, 6
Kugeln Magnetkugeln
6x1,5 ml Position magnetic separator
  • Magnetic separation
  • Six 1,5 ml microcentrifuge tubes
A magnetic separation unit designed to accommodate up to six 1,5 ml microcentrifuge tubes in order to separate magnetic particles contained in those tubes. Optimal working volume is up to 1 ml.
Magnetic separator unit can be used to hold, and separate magnetic particles.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge