Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Respirators, industrial, PX5 PAPR Series

Lieferant: RPB SAFETY

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

03-801-FR-EUUK 03-891 04-854
RPBS03-801-FR-EUUKEA 0 EUR InStock
RPBS03-801-FR-EUUK RPBS03-891 RPBS04-854
Respirators, industrial, PX5 PAPR Series
Atemschutzmasken Atemschutz-Systeme
The RPB PX5 is a battery powered air purifying respirator fan unit which draws in contaminated air from the wearer's immediate vicinity, passing it through internal filters, and supplying the filtered air to an approved RPB loose fitting respirator head top.

  • Backpack option available for applications that require a greater range of movement and work across varied angles
  • With a completely sealed housing, the PX5 has been certified to IP 65 rating in use and certified to IP 67 rating with cleaning kit in place
  • Filter door cannot mount to the unit unless a filter is fixed in place
  • HEPA filter removes up to 99,97% of particulates down to 0,12 µm
  • Gas filter protects against organic vapor exposure (replace pre-filter daily to extend the life of main filter)
  • Visual, audible, and vibrating alarms for low battery level, or when airflow falls below the minimum requirements
  • Long-life lithium-ion battery is protected from moisture when decontaminating and will not disconnect or bump off while operating
  • Operating temperature: −20 to +50 °C
  • Noise rating: speed 1: 57 dBA, speed 2: 59 dBA, speed 3: 62 dBA
  • Flow rate: speed 1: 170 L/min, speed 2: 210 L/min, speed 3: 240 L/min

Weighing just 1,16 kg with HEPA filter fitted (1,65 to 1,7 kg with gas filter fitted), the sleek, lightweight, compact design maximizes comfort. PX5 unit attaches to a flexi-belt system (fitted with the decon easy clean belt) via an intuitive vertical mount lock system (easily removes for quick and simple decontamination).

Deigned to be as compact and comfortable as possible, the Easy clean PX5 Starter kit is perfect for use within infection control, and cleanroom environments, where easily and effectively cleaning the system is of utmost importance. Depending on what type of contaminant users require protection from, the PX5 has various filter options, including gas and multi filters. The PX5 also has built-in audible, visual, and physical alarms to warn the wearer when their filter needs to be changed, or when there is only 5% of the battery charge remaining.

The unit is IP 65 (in use) and IP 67 rated.

Zertifizierungen: EN 12941:1998+A2:2008 TH3/2 P R SL.

Lieferumfang: Includes 1×PX5 PAPR, 1×decon easy clean belt, 1×particulate filter, 1×pack of 10 prefilters, 1×lithium ion rechargeable battery, 1×charging bank with EUUK plugs, and 1×flow tester. Note: head top and breathing tube sold separately.

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge