Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Respirators with FR chin seal, Z-Link Series

Lieferant: RPB SAFETY

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

16-010-31-CE 16-670 16-810 16-811 16-531 16-522
RPBS16-010-31-CEEA 0 EUR InStock
RPBS16-010-31-CE RPBS16-670 RPBS16-810 RPBS16-811 RPBS16-531 RPBS16-522
Respirators with FR chin seal, Z-Link Series
Atemschutzmasken Atemschutz-Systeme
The RPB Z-link respirator with FR chin seal, is the world's most comfortable (fully padded) and versatile industry respirator. Providing the highest respiratory protection available (TH3), the Z-Link system also provides hard hat protection, eye and face protection and hearing protection.

  • Visor hinges open for easy communication with team (outside of respiratory hazards)
  • Air flow and distribution is fully adjustable with the air deflector system to suit personal preferences
  • Fixed jaw ensures secure cape attachment
  • Vision-Link™ light mount point
  • ANSI Z89.1 certified head protection (EN 397)
  • Padded interior with adjustable ratchet system ensures a perfect customized fit that supports movement, increasing comfort and productivity
  • Three cape materials available: Tychem® 2000, Tychem® 4000, or Zytec® FR
  • Operating temperature: −10 to +60 °C
  • Noise rating: NRR 25 (side pads), NRR 41 (ear defenders and earplugs)

Optimum vision and high velocity impact protection with large optically-clear safety lens (replaceable spare part). Choice of 5 antifog and scratch resistant ANSI Z87+ (EN 166) lenses: premium clear, green tint (shade 5), yellow tint, black tint and mirror tint.

The Z-Link is designed to be able to perform all tasks within a business, including general RPE, welding, grinding, painting/coating, and now high heat foundry works. This allows companies to standardize systems and reduce alternate system and brands spare part stocking and usage.

To achieve this, the Z-Link offers a wide range of market leading out of the box accessory items to ensure user comfort, safety and productivity. These include:
• Clip on weld visor with 1:1:1 ADF (16-670)
• Chin mounted helmet light (16-901)
• In-helmet communication system (16-922)
• Quiet-link ear defenders (18-533)
• Comfort fit side pads (as standard)
• IR5 visors (16-810-IR5, 16-675-IR5)
• High heat foundry kits, including gold visors (16-674, 16-744, 04-955, 16-675-GT)

Zertifizierungen: EN 12941:1998+A2:2008 TH3/2 P R SL, EN 397:2012+A1:2012, EN 166:2001 Class 1BT 9 3.

Lieferumfang: Includes PX5 PAPR breathing tube. PAPR sold separately.

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge