Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-BrdU Mouse Monoclonal Antibody

Lieferant: Cytiva

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

RPN202
RPN202EA 1074 EUR InStock
RPN202
Anti-BrdU Mouse Monoclonal Antibody
Antikörper
Anti-Bromodeoxyuridine (Anti-BRdU) antibody is supplied in the same formulation as the monoclonal antibody to BrdU in the Cell Proliferation Kit. The antibody/nuclease mixture is compatible with detection systems that incorporate fluorescently labelled antibodies and biotin-streptavidin detection reagents.

  • No need for chemical denaturation
  • Detection while maintaining cell morphology
  • Double immunocytochemical staining studies can be perfromed

Used in conjunction with RPN20, the reagent allows extended use of the kit where additional amounts of labeling reagent are required, such as in vivo studies.

The antibody contains a nuclease to produce single stranded DNA without the need for chemical denaturation. Cellular morphology is preserved allowing multiple labeling experiments to be carried out. The high-contrast stain produced has sensitivity comparable to autoradiographic methods.

Type: Primary
Antigen: BrdU (Bromodeoxyuridine)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Chemical

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge