Western Blotting-Blockierungsreagenz, Amersham ECL™
Lieferant: GE HEALTHCARE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
RPN2125
RPN2125EA 89.2 EUR InStock
RPN2125
Western Blotting-Blockierungsreagenz, Amersham ECL™
Elektrophorese-Reagenzien Western Blotting Reagents
Amersham ECL Blocking Agent is for blocking nitrocellulose and PVDF membranes to be used with Amersham ECL, Amersham ECL Prime and Amersham ECL Select Western Blotting Detection Reagents
Can be used with the Amersham ECL Western blotting detection family. This blocking agent is intended for blocking nitrocellulose and PVDF membranes used with ECL and ECL Plus Western Blotting Detection Reagents.
Lieferumfang: Jede Packung enthält 40 g des Membran-Blockierungsreagenz, ausreichend für mindestens 20 Miniblots.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge