Rinse solution no. 2, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
Achtung
RS2-900ML
RS2-900MLEA 90.4 EUR InStock
RS2-900ML
Rinse solution no. 2, Sigma-Aldrich®
Rinse solution no. 2
Intended for use as a buffer with various Romanowky type staining procedures, specifically those performed on the Hematek. RS2 is a component of Sigma products WGHT, WSHT and WSHT-S.
IVDLogo
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge