Donor horse serum
Lieferant: Biowest
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
S0900-100 S0900-500 S090H-100 S090H-500 S0900-1000 S090Y-100 S090Y-500 S090G-100
S0900-100EA 20.8 EUR InStock
S0900-100 S0900-500 S090H-100 S090H-500 BWSTS0900-1000 S090Y-100 S090Y-500 S090G-100
Donor horse serum
Seren
Donor horse serum is aseptically collected from herds of healthy animals inspected periodically by a licensed veterinarian. Applications for donor horse serum include mycoplasma contamination tests carried out on media, blocking agent and controls in immunoassays, culture of PC12 cells when combined with FBS.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge