Transfer pipettes, general purpose, Samco™
Lieferant: SAMCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
202-1S 202-20S 202 273 273-1S 336-1S 335-11
SAMC202-1SEA 0 EUR InStock
SAMC202-1S SAMC202-20S SAMC202 SAMC273 SAMC273-1S SAMC336-1S SAMC335-11
Transfer pipettes, general purpose, Samco™
Pipetten Pasteur- und Transfer-Pipetten
These transfer pipettes are made of shatterproof, non toxic polyethylene. They have a built-in bulb that eliminates the need for plugging, packaging and autoclaving.
  • Low-affinity surface reduces loss of cells and valuable proteins due to binding
  • Pipettes can be frozen in liquid nitrogen, or heat sealed for storage
  • Non graduated
Ideal for tissue culture applications, routing sampling of serum and plasma, blood culture, urine analysis.
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge