Transfer pipettes, graduated, Samco™
Lieferant: SAMCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
212 212-1S 212-20S 222 222-1S 274 222-20S 225 225-1S 225-20S
SAMC212EA 0 EUR InStock
SAMC212 SAMC212-1S SAMC212-20S SAMC222 SAMC222-1S SAMC274 SAMC222-20S SAMC225 SAMC225-1S SAMC225-20S
Transfer pipettes, graduated, Samco™
Pipetten Pasteur- und Transfer-Pipetten
These graduated pipettes feature a built-in pipette bulb that eliminates need for plugging, packaging, autoclaving while preventing cross-contamination.
  • Made of shatterproof, non toxic polyethylene
  • Low-affinity surface reduces loss of cells and valuable proteins due to binding
  • Can be frozen in liquid nitrogen, or heat sealed for storage
  • Accurately measure more than one volume
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge