Transfer pipettes, narrow stem, Samco™
Lieferant: SAMCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
241 242 251-1S 251-20S 251 252
SAMC241EA 0 EUR InStock
SAMC241 SAMC242 SAMC251-1S SAMC251-20S SAMC251 SAMC252
Transfer pipettes, narrow stem, Samco™
Pipetten Pasteur- und Transfer-Pipetten
These pipettes made of shatterproof, non toxic PE prevent cross-contamination and are used for blood bank applications such as sedimentation procedures.
  • Low-affinity surface reduces loss of cells and valuable proteins due to binding
  • Can be frozen in liquid nitrogen, or heat sealed for storage
  • No need for plugging, packaging and autoclaving
The single-unit design features a built-in pipette bulb, and a flexible stem bends to sample from narrow or small tubes.
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge