Transfer pipettes, Samco™ Extra Long
Lieferant: SAMCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
262-1S 262-20S 262-10S 263 263-1S
SAMC262-1SEA 0 EUR InStock
SAMC262-1S SAMC262-20S SAMC262-10S SAMC263 SAMC263-1S
Transfer pipettes, Samco™ Extra Long
Pipetten Pasteur- und Transfer-Pipetten
These extra long PE pipettes are ideal for safe and easy sampling from large containers.
  • Excellent for tissue culture applications
  • General purpose pipettes are ideal for routine sampling of serum and plasma
  • Well suited for blood culture and urine analysis
Perfect for tissue culture applications, these pipettes are designed with a built-in pipette bulb that eliminates the need for plugging, packaging, autoclaving and prevents cross-contamination.
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge