Exact volume transfer pipettes, Samco™, Thermo Scientific
Lieferant: SAMCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
787NL 966NL 191NL 783NL 784NL 785NL 786NL 788NL 930NL 941NL 967NL
SAMC787NLEA 0 EUR InStock
SAMC787NL SAMC966NL SAMC191NL SAMC783NL SAMC784NL SAMC785NL SAMC786NL SAMC788NL SAMC930NL SAMC941NL SAMC967NL
Exact volume transfer pipettes, Samco™, Thermo Scientific
Pipetten Pasteur- und Transfer-Pipetten
The exact volume transfer pipettes are used for dispensing reagents of small samples.
  • Polyethylene
  • Non sterile
  • Shatterproof
  • Ideal for home and POC (Point of Care) testing
  • OEM applications
Their low-affinity surface prevents proteins binding in research applications. They can be frozen in liquid nitrogen, or heat sealed for storage. Useful in clinical laboratories. Custom design options available to meet your specifications.
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge