Sample spoons, USP Class VI, double-bagged, ULTRA
Lieferant: Sampling Systems
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
9088H-03S 9088H-10S
SAMP9088H-03SEA 298 EUR InStock
SAMP9088H-03S SAMP9088H-10S
Sample spoons, USP Class VI, double-bagged, ULTRA
Löffel
These sterile HDPE sampling spoons are great for taking powder and liquid samples. They are commonly used for taking sterile samples from tubs, bags, containers, bins etc.
  • Double-bagged
  • Produced in Class 100,000 cleanroom, packed in Class 10,000 cleanroom
The spoons are designed to be used straight from the bag. The double bagging means that the outer bag can be removed whilst entering the cleanroom, ensuring the cleanliness inside the cleanroom or manufacturing area.
Zertifizierungen: USP Class VI Medical grade polymer is FDA and EC compliant, tested to USP661. Certified BSE/TSE free.
Verpackung: Supplied in a lined box.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge