Powder funnels, USP Class VI, double-bagged, ULTRA
Lieferant: Sampling Systems
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
9111H-200S
SAMP9111H-200SEA 205 EUR InStock
SAMP9111H-200S
Powder funnels, USP Class VI, double-bagged, ULTRA
Trichter Pulvertrichter
The SteriWare Ultra range is the premium range from Sampling Systems. These powder funnels are double bagged and are made from high quality USP class VI HDPE. A box liner is also used giving a third layer of protection.
  • Double-bagged
  • Produced in class 100,000 cleanroom, packed in Class 10,000 cleanroom
  • Crevice free with smooth finish
  • Angle of the cone is designed to give optimum product flow
These sterile funnels are designed to be used straight from the bag. The double bagging means that the outer bag can be removed whilst entering the cleanroom, ensuring the cleanliness inside the cleanroom or manufacturing area.
This range is ideal for demanding applications such as GMP, vaccine and cleanroom manufacturing. The funnels have a 200 mm diameter.
Zertifizierungen: USP Class VI Medical grade polymer is FDA and EC compliant, tested to USP661.
Verpackung: Supplied in a lined box.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge