Chemical Spoon and Spatulas
Lieferant: Sampling Systems
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
A625-150 A625-280 A625-320
SAMPA625-150EA 10.1 EUR InStock
SAMPA625-150 SAMPA625-280 SAMPA625-320
Chemical Spoon and Spatulas
Löffel
Versatile spoon and spatula for sampling powder and liquid products.
  • Available in a range of sizes
  • Heavy duty design for comfortable use
  • Can be used as both a spoon and a spatula
  • Perfect for measuring, sampling and dosing powders and liquids
This versatile product has a robust design and is made from 304 stainless steel. It can be used as both a spoon and a spatula allowing greater versatility in the sampling of both powder and liquid products. There are no sharp edges or crevices and are ideal for use in food, pharma and chemical sectors.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge