Stainless Steel Bottles, GL45
Lieferant: Sampling Systems
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
A886-100 A886-1000 A886-500
SAMPA886-100EA 1090 EUR InStock
SAMPA886-100 SAMPA886-1000 SAMPA886-500
Stainless Steel Bottles, GL45
Flaschen Metallflaschen
These selection of GL45 stainless bottles are suitable a wide range of laboratory and production applications.
  • Made from corrosion resistant 316L stainless steel
  • Includes a widely used GL45 bottle thread
  • Featuring an airtight screw cap with a PTFE sealing ring
The range of GL45 stainless bottles are perfect for storing liquids and are used across a wide range of industries. The GL45 thread, cap and PTFE sealing ring provide a hermetic seal protecting the contents from external factors.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge