Sterilising autoclaves, LAS
Lieferant: SANOCLAV WOLF
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
01018
SANC01018EA 2265.49 EUR InStock
SANC01018
Sterilising autoclaves, LAS
Sterilisatoren Autoklaven (Dampfsterilisatoren)
Lid and chamber made of stainless steel, suitable for the sterilisation in tightened steam and water.
  • With whistle and safety valve
  • Includes steam pressure regulation valve
  • Without controller and with temperature and pressure gauge
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge