Analytical laboratory balances, Cubis® II MCA
Lieferant: Sartorius Balances
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
MCA524S2S00U99 MCA524P2S00U99 MCA324S2S00U99 MCA324P2S00U99 MCA224S2S00U99 MCA124S2S00U99 MCA524S2S00A99 MCA524P2S00A99 MCA324S2S00A99 MCA324P2S00A99 MCA224S2S00A99 MCA124S2S00A99 MCA524S2S00I99 MCA524P2S00I99 MCA324S2S00I99 MCA324P2S00I99 MCA224S2S00I99 MCA124S2S00I99 MCA324P2S00-I MCA524S2S00-U MCA524P2S00-U MCA324S2S00-U MCA324P2S00-U MCA224S2S00-U MCA124S2S00-U MCA524S2S00-A MCA524P2S00-A MCA324S2S00-A MCA324P2S00-A MCA124S2S00-A MCA524S2S00-I MCA524P2S00-I MCA324S2S00-I MCA224S2S00-I MCA124S2S00-I MCA224S-2S00-A
SARBMCA524S2S00U99EA 15340 EUR InStock
SARBMCA524S2S00U99 SARBMCA524P2S00U99 SARBMCA324S2S00U99 SARBMCA324P2S00U99 SARBMCA224S2S00U99 SARBMCA124S2S00U99 SARBMCA524S2S00A99 SARBMCA524P2S00A99 SARBMCA324S2S00A99 SARBMCA324P2S00A99 SARBMCA224S2S00A99 SARBMCA124S2S00A99 SARBMCA524S2S00I99 SARBMCA524P2S00I99 SARBMCA324S2S00I99 SARBMCA324P2S00I99 SARBMCA224S2S00I99 SARBMCA124S2S00I99 SARBMCA324P2S00-I SARBMCA524S2S00-U SARBMCA524P2S00-U SARBMCA324S2S00-U SARBMCA324P2S00-U SARBMCA224S2S00-U SARBMCA124S2S00-U SARBMCA524S2S00-A SARBMCA524P2S00-A SARBMCA324S2S00-A SARBMCA324P2S00-A SARBMCA124S2S00-A SARBMCA524S2S00-I SARBMCA524P2S00-I SARBMCA324S2S00-I SARBMCA224S2S00-I SARBMCA124S2S00-I 611-5264
Analytical laboratory balances, Cubis® II MCA
Waagen
Cubis® II MCA analytical balances with advanced user interface offer maximum weighing capacity between 120 and 520 g in combination with readability of 0.1 mg.
  • Modular premium balances with thousands of hardware and software configurations and adaptable software
  • Seamless connectivity
  • Long-term flexibility
  • Submission-ready data
  • Governance through user management
  • Integration of SOPs
  • Intuitive applications
  • Status centre notifications
Weighing in a regulated environment can be complex, with stringent compliance directives and controls required to deliver the accuracy and reliability of results required by the pharmaceutical industry. One of the technologies helping scientists to achieve this level of accuracy is the Cubis® II balance series from Sartorius.
All weighing modules of the Cubis® II balance are designed for intuitive operation, further aided by intelligent diagnostic systems. These design elements guarantee a higher degree of repeatability for different workflows while lowering the probability of human error during measurement. It offers modern user interfaces, pharmaceutical and GxP compliance, including data handling, data integrity and connectivity, ergonomic sample handling, easy process integration, and unlimited communication at the highest level of accuracy and precision.
These MCA models with Advanced User Interface have a high resolution colour touch screen plus holders for ergonomic sample handling and automatic motorised levelling. The balances feature innovative draft shield technology, gesture sensor and climate monitoring. With complete traceability the balances help achieve 21 CFR Part 11 and EU Annex 11 compliance.
Choice of draft shield denoted by model name suffix: Manual glass doors with large draft shield (U); motorised draft shield (A) or motorised with built-in de-ioniser (I) for effective removal of static charges.
Bestellinformation: There are different optional software packages, each contains applications or functions to enhance and customise the balance for the users' workflows. The all inclusive QApps package (denoted by QP99) includes all four of these packages: Pharma; Advanced Applications; Utilities and Connectivity.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge