Cubis II MCA electronic high capacity micro balance with large high end 7 color TFT touch display in 16:9 format with graphical user interface. Actual scale division (d) 0.001 mg, nominal maximum load (Max) 61 g. Automatic motorized glass draft shie 1 * 1 ST
Lieferant: SARTORIUS BALANCES
Cubis II MCA electronic high capacity micro balance with large high end 7 color TFT touch display in 16:9 format with graphical user interface. Actual scale division (d) 0.001 mg, nominal maximum load (Max) 61 g. Automatic motorized glass draft shie 1 * 1 ST
New Product
PRODUKT
Erweitert / Kompakt
Cubis II MCA electronic high capacity micro balance with large high end 7 color TFT touch display in 16:9 format with graphical user interface. Actual scale division (d) 0.001 mg, nominal maximum load (Max) 61 g. Automatic motorized glass draft shie 1 * 1 ST SARBMCA66S3CEUDQP1 1 * 1 ST
New Product
Hersteller-Artikelnummer:: MCA66S3CEUDQP1
Lokale Artikelnummer:: SARBMCA66S3CEUDQP1
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder