High Resolution OEM Weigh Cell
Lieferant: Sartorius Balances
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
WZA245-NC
SARBWZA245-NCEA 17250 EUR InStock
SARBWZA245-NC
High Resolution OEM Weigh Cell
Waagen
The WZA-NC series provide a readability of 1/10 µg for the weighing capacity of 20/240 g. The weighing system
as well the electronic are designed for space sensitive applications in stainless steel housings.
  • Load receptor with overload protection
  • Force compensation technology system with a resolution of 1/10 µg
  • Superb electronics for 24 Mio. step resolution
  • Internal calibration weight for easy checks in automated environments
  • Possibility for under floor weighing
  • Fast warm-up by separation of weighing system and electronics
  • USB|RS232C interface for configuration and data transfer
The weigh cell consist of two parts:
A monolithic weighing system in stainless steel IP44 housing.
Stainless steel electronic box, all connection via the front.
Features optional PC-software with menu setting, weigh data display and internal calibration functions
Applications can be found in weighing of samples in automated system, manual pipette calibration and in automated liquid control, e.g. LCD productions.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge