Kalibriergewichte, Klasse F1, Proof Line
Lieferant: Sartorius Balances
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
YCW413-00 YCW513-02 YCW413-02 YCW023-02 YCW153-00 YCW153-02 YCW213-00 YCW213-02 YCW023-00 YCW013-00 YCW013-02 YCW223-00 YCW423-00 YCW553-00 YCW353-00 YCW313-00 YCW323-02 YCW723-02 YCW653-00 YCW653-02 YCW713-02 YCW223-02 YCW123-00 YCW053-00 YCW113-02 YCW123-02 YCW313-02 YCW353-02 YCW513-00 YCW523-00 YCW723-00 YCW113-00 YCW253-00 YCW053-02 YCW253-02 YCW613-02 YCW423-02 YCW523-02 YCW623-02 YCW453-02 YCW553-02 YCW613-00 YCW323-00 YCW623-00 YCW453-00 YCW713-00 YCW313-AC-02 YCW353-AC-02 YCW353-AC-00 YCW313-AC-00 YCW323-AC-02 YCW323-AC-00 YCW623-AC-02 YCW623-AC-00 YCW653-AC-00 YCW613-AC-02 YCW613-AC-00 YCW713-AC-02 YCW713-AC-00 YCW653-AC-02 YCW553-AC-00 YCW423-AC-00 YCW453-AC-02 YCW453-AC-00 YCW423-AC-02 YCW413-AC-02 YCW413-AC-00 YCW523-AC-02 YCW553-AC-02 YCW513-AC-02 YCW513-AC-00 YCW523-AC-00
SARBYCW413-00EA 93.6 EUR InStock
SARBYCW413-00 SARBYCW513-02 SARBYCW413-02 SARBYCW023-02 SARBYCW153-00 SARBYCW153-02 SARBYCW213-00 SARBYCW213-02 SARBYCW023-00 SARBYCW013-00 SARBYCW013-02 SARBYCW223-00 SARBYCW423-00 SARBYCW553-00 SARBYCW353-00 SARBYCW313-00 SARBYCW323-02 SARBYCW723-02 SARBYCW653-00 SARBYCW653-02 SARBYCW713-02 SARBYCW223-02 SARBYCW123-00 SARBYCW053-00 SARBYCW113-02 SARBYCW123-02 SARBYCW313-02 SARBYCW353-02 SARBYCW513-00 SARBYCW523-00 SARBYCW723-00 SARBYCW113-00 SARBYCW253-00 SARBYCW053-02 SARBYCW253-02 SARBYCW613-02 SARBYCW423-02 SARBYCW523-02 SARBYCW623-02 SARBYCW453-02 SARBYCW553-02 611-1707 SARBYCW323-00 611-1708 SARBYCW453-00 SARBYCW713-00 SARBYCW313-AC-02 SARBYCW353-AC-02 SARBYCW353-AC-00 SARBYCW313-AC-00 SARBYCW323-AC-02 SARBYCW323-AC-00 SARBYCW623-AC-02 SARBYCW623-AC-00 SARBYCW653-AC-00 SARBYCW613-AC-02 SARBYCW613-AC-00 SARBYCW713-AC-02 SARBYCW713-AC-00 SARBYCW653-AC-02 SARBYCW553-AC-00 SARBYCW423-AC-00 SARBYCW453-AC-02 SARBYCW453-AC-00 SARBYCW423-AC-02 SARBYCW413-AC-02 SARBYCW413-AC-00 SARBYCW523-AC-02 SARBYCW553-AC-02 SARBYCW513-AC-02 SARBYCW513-AC-00 SARBYCW523-AC-00
Kalibriergewichte, Klasse F1, Proof Line
Gewichte Kalibriergewichte
Proof Line weights are high quality working standards for everyday use. These weights are made of high quality material with polished or precision lathed surfaces.
  • 0 to 5 mg leaf weights: Aluminium
  • 10 to 500 mg leaf weights: Nickel silver
  • 1 g to 10 Kg knob weights: Stainless steel
Zertifizierungen: Compliance with OIML R111-1.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge