Filtration capsules, Sartopore® Platinum
Lieferant: Sartorius
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
5495307H8-OO-A 5495307H9-OO-A
SARP5495307H8-OO-AEA 1140 EUR InStock
SARP5495307H8-OO-A SARP5495307H9-OO-A
Filtration capsules, Sartopore® Platinum
Filter Membranfilterkapseln
Sartopore® Platinum sterilising grade MidiCaps® are self-contained sterile filter units designed for pharmaceutical and biotechnology industry. These filters contain a unique heterogeneous double layer of hydrophilised polyethersulfone membranes and are specially designed to meet the requirements for filtration of a broad range of pharmaceutical products.
  • Ready to use
  • Ideal for scale-up
  • Compatible with a pH range from 1 to 14 pH
  • Can be easily wetted with lowest flushing volume
PES membrane; support fleece, core and end caps: PP; O-rings: silicone; housing: PP
Sartopore® Platinum Midicaps® are designed, developed and manufactured in accordance with a ISO 9001 certified Quality Management System. A Validation Guide is available for compliance with regulatory requirements.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge