Sartobind® Filtration Capsules
Lieferant: Sartorius
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
96HICP42EUC11A 92IEXQ42D4-OOA 92STPA42DD-11D 92IEXQ42D4-SSA 92IEXS42D4-OOA 92IEXS42D4-SSA 92IEXQ42DD-11D 92HICP42DD-11D
SARP96HICP42EUC11AEA 690 EUR InStock
SARP96HICP42EUC11A SARP92IEXQ42D4-OOA SARP92STPA42DD-11D SARP92IEXQ42D4-SSA SARP92IEXS42D4-OOA SARP92IEXS42D4-SSA SARP92IEXQ42DD-11D SARP92HICP42DD-11D
Sartobind® Filtration Capsules
Filter Membranfilterkapseln
Ready to use disposable ion exchange capsules for capturing and polishing in biomanufacturing. Ideal for applications such as the removal of DNA, viruses, endotoxins and protein contaminants.
  • Simple handling
  • Fast operation
  • Less buffer consumption
Stabilised reinforced cellulose.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge