Deep well V-bottom plates, 24-well
Lieferant: Sartorius
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
A-0038
SARPA-0038EA 780 EUR InStock
SARPA-0038
Deep well V-bottom plates, 24-well
Mikrotiterplatten Deep-Well-Mikroplatten
These 24-well deep well plates can be used to hold samples harvested from bioreactors. They can also be used to hold reagents and feeds on the storage deck, before being added into the bioreactor.
  • Irradiated
Lieferumfang: Pack contains 25 sterile plates with lids, individually wrapped.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge