Blood collecting tubes with serum gel (clotting activator), S-Monovette®
Lieferant: SARSTEDT
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
06.1667.001 04.1935 01.1602.001 03.1524.001 04.1905.001 04.1935.001 02.1388.001 01.1602.00 02.1388 03.1524 04.1905 04.1905.001-50 04.1935.001-50
SARS06.1667.001EA 204 EUR InStock
SARS06.1667.001 SARS04.1935 SARS01.1602.001 SARS03.1524.001 SARS04.1905.001 SARS04.1935.001 SARS02.1388.001 SARS01.1602.00 SARS02.1388 SARS03.1524 SARS04.1905 SARS04.1905.001-50 SARS04.1935.001-50
Blood collecting tubes with serum gel (clotting activator), S-Monovette®
Klinisches Diagnosezubehör Klinische Probenentnahme und Probenvorbereitung Entnahme und Vorbereitung von Blutproben
S-Monovette® is a blood collection system that combines the aspiration and the vacuum techniques.
  • Aspiration technique for routine blood collection
  • Vacuum technique is always available
  • For all ages, from young to old
Suitable for all vein conditions and achieves a sample quality for high quality results.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge