Pipette tips
Lieferant: SARSTEDT
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
70.3031 70.3030.100 70.3050 70.3050.200 70.3010 70.3010.200 70.3010.100 70.760.201 70.3050.205 70.3040 70.1186.100 3021 70.1116.200
SARS70.3031EA 139 EUR InStock
SARS70.3031 SARS70.3030.100 SARS70.3050 SARS70.3050.200 SARS70.3010 SARS70.3010.200 SARS70.3010.100 SARS70.760.201 SARS70.3050.205 SARS70.3040 SARS70.1186.100 SARS3021 SARS70.1116.200
Pipette tips
Pipettenspitzen
These high quality tips are suitable for the most common pipettes.
  • Sterile tips are DNase/RNase- and ATP-free, and non pyrogenic
  • Available loose in bags, packed in racks or in Tip StackPack reload systems
Tip StackPack: Tips are preloaded in reusable, stacked 96-tip trays. The trays are designed to safely cover each individual 96-tip layer, shielding the preloaded tips in the stack like a protective screen.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge