Microcentrifuge tubes, SafeSeal
Lieferant: SARSTEDT
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
72.706 72.706.200 72.695.001 72.695.002 72.695.004 72.695.005 72.695.006 72.695.007 72.695.200 72.704 72.704.002 72.706.002 72.704.004 72.704.005 72.704.006 72.695.500 72.706.400
SARS72.706EA 0 EUR InStock
SARS72.706 SARS72.706.200 SARS72.695.001 SARS72.695.002 SARS72.695.004 SARS72.695.005 SARS72.695.006 SARS72.695.007 SARS72.695.200 SARS72.704 SARS72.704.002 SARS72.706.002 SARS72.704.004 SARS72.704.005 SARS72.704.006 SARS72.695.500 SARS72.706.400
Microcentrifuge tubes, SafeSeal
Röhrchen Mikroröhrchen
PP, translucent.
  • Wide hinge for easy and precise closure
  • Large writing space on lid, graduated
  • Centrifugation to 30000×g (2,0 ml tubes: to 25000×g)
For transport, storage and sample processing in the laboratory. DNA-/RNase-free tubes and screw caps for the storage of high-grade synthesised oligonucleotides, enzymes, buffers etc. Retaining cams ensure a safe fit also under thermal strain.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge