Micro tubes with thread
Lieferant: SARSTEDT
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
72.693.465 72.730.217
SARS72.693.465EA 223 EUR InStock
SARS72.693.465 SARS72.730.217
Micro tubes with thread
Röhrchen Mikroröhrchen
PP, translucent with PP screw cap.
  • With printed label
  • Withstand centrifugation up to 20000×g
  • Attached, not screwed-on cap
  • Sterile and free of DNA, DNase/RNase, PCR inhibitors, ATP and pyrogens/endotoxins
With or without skirted base.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge