Deep well plates, 96-well, PS, MegaBlock®
Lieferant: SARSTEDT
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
82.1970.002
SARS82.1970.002EA 173 EUR InStock
SARS82.1970.002
Deep well plates, 96-well, PS, MegaBlock®
Mikrotiterplatten Deep-Well-Mikroplatten
PS. The 96-well, 1,2 ml MegaBlock® meets all important requirements for automated sample processing or storing large quantities of backup samples.
  • Each well 100% leakproof tested
  • Ideal for longterm storage of blood samples
  • Transparent for superior visual control
  • Round bottom wells
  • Alphanumeric coding for easy identification
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge