Blood collection needles
Lieferant: SARSTEDT
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
14.1205 85.1162 14.1112 85.1160 85.1637.035 85.1160.200 85.1162.200 85.1440.200 85.1637.205 85.1637.235 85.1638.200 85.1638.205 85.1638.235 85.1640.205 85.1640.235 85.1642.205 14.1205.100
SARS14.1205EA 293 EUR InStock
SARS14.1205 SARS85.1162 SARS14.1112 SARS85.1160 SARS85.1637.035 SARS85.1160.200 SARS85.1162.200 SARS85.1440.200 SARS85.1637.205 SARS85.1637.235 SARS85.1638.200 SARS85.1638.205 SARS85.1638.235 SARS85.1640.205 SARS85.1640.235 SARS85.1642.205 SARS14.1205.100
Blood collection needles
Injektionsnadeln
The needle protection on the safety-needles and Safety-Multifly® needles enables the safe termination of blood collection by locking the needle in the needle protector.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge