Multi-tube racks, S-Monovette®
Lieferant: SARSTEDT
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
93.837 93.826 93.850 93.841.100 93.1431.001 93.838 93.848.100 93.894 93.1097.100 93.1431 93.48.011 93.827 93.827.013 93.838.013 93.844.100 93.846.100 93.849.100 93.850.001 93.868 93.870 93.893.100 93.1428
SARS93.837EA 52.4 EUR InStock
SARS93.837 SARS93.826 SARS93.850 SARS93.841.100 SARS93.1431.001 SARS93.838 SARS93.848.100 SARS93.894 SARS93.1097.100 SARS93.1431 SARS93.48.011 SARS93.827 SARS93.827.013 SARS93.838.013 SARS93.844.100 SARS93.846.100 SARS93.849.100 SARS93.850.001 SARS93.868 SARS93.870 SARS93.893.100 SARS93.1428
Multi-tube racks, S-Monovette®
Gestelle Reagenzglasgestelle
Made from unbreakable, transparent plastic and can be easily disassembled for cleaning.
  • A wide range with a variety of diameters
  • High-grade and resistant material that is durable
  • Available in different colours
These universal compact block rack are ideal for space-saving and systematic sample storage.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge