Transport Boxes
Lieferant: SARSTEDT
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
95.1715 95.1717 95.1717.003 95.1717.011
SARS95.1715EA 0 EUR InStock
SARS95.1715 SARS95.1717 SARS95.1717.003 SARS95.1717.011
Transport Boxes
Boxen Transportboxen
Transport case housing is manufactured from durable polyethylene and offers optimum protection for primary receptacles during transport.
  • Economical, flexible
  • Central one-hand closure
  • Lateral tabs for easy attachment of straps or restraints
  • Transparent window label for easy identification
Transport cases are suitable for transportation of diagnostic samples and blood products.
Zertifizierungen: Complies with the requirement of P650 packaging instructions for class UN3373 of the ADR and RID.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge