Kryoboxen
Lieferant: SARSTEDT
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
95.64.922 95.64.923 95.64.924 95.64.981 95.64.982 95.64.997
SARS95.64.922EA 649 EUR InStock
SARS95.64.922 SARS95.64.923 SARS95.64.924 SARS95.64.981 SARS95.64.982 SARS95.64.997
Kryoboxen
Boxen Kryoboxen
Storage boxes, moisture-resistant laminated carboard material with lid and compartmentalised insert.
  • Space saving and organised sample storage
  • Safe and light-protected storage
Suitable for micro tubes, PCR tubes and centrifuge tubes.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge