Syringe filters, Minisart® Acticosart
Lieferant: Sartorius
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
17840-Q
SART17840-QEA 2510 EUR InStock
SART17840-Q
Syringe filters, Minisart® Acticosart
Filter Spritzenvorsatzfilter
The Minisart® Acticosart syringe filter with chemically inert hydrophobic PTFE is suitable for venting as well as filtration of gases with a pH of 3 to 12.
  • MBS housing suitable for glueing
  • PVC-free
  • 100% optical integrity test
  • Colour coded MBS housing for easy pore identification
  • Superior flow rate
Due to its hydrophobic nature the PTFE membrane functions as a water barrier and does not become wetted by aerosols. This keeps the hydrophobic venting filter permeable for gas or air. The active carbon filling adsorbs molecules. The housing of the filter is made of MBS (Methacrylate butadiene styrene) which has a limited compatibility with alcohols but is free of phthalates and passes the plastic class VI test.
Verpackung: Supplied in bulk packs.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge