Filter papers, non woven
Lieferant: Sartorius
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
FT-3-01304-025
SARTFT-3-01304-025EA 176 EUR InStock
SARTFT-3-01304-025
Filter papers, non woven
Filter Filterpapiere Qualitatives Filterpapier
These non woven grades are made of rayon. Ideal for the filtration or prefiltration of viscous solutions containing particles visible with the naked eye.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge