Qualitative filter papers, technical, smooth, grade 3 S/h
Lieferant: Sartorius
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
FT-3-307-150 FT-3-307-070 FT-3-307-240 FT-3-307-185 FT-3-307-110 FT-3-307-100 FT-3-307-055 FT-3-307-090 FT-3-307-125 FT-3-307-270 FT2-307-580580 FT2-307-460570
SARTFT-3-307-150EA 20.3 EUR InStock
SARTFT-3-307-150 SARTFT-3-307-070 SARTFT-3-307-240 SARTFT-3-307-185 SARTFT-3-307-110 SARTFT-3-307-100 SARTFT-3-307-055 SARTFT-3-307-090 SARTFT-3-307-125 SARTFT-3-307-270 SARTFT2-307-580580 SARTFT2-307-460570
Qualitative filter papers, technical, smooth, grade 3 S/h
Filter Filterpapiere Qualitatives Filterpapier
These smooth filter papers are made of refined pulp and linters with more than 95% alpha-cellulose content. The ash content of these filter papers is between 0,1 and 0,15%.
They are used for routine analyses like clarification, determination of substances, but also as discs with a centre hole for technical applications. Grades with a wet burst resistance >30 kPa are referred to as wet-strengthened and are therefore suitable for pressure or vacuum filtration.
Medium fast to slow rate of filtration, narrow-pore; retains fine particles. Ideal for difficult-to-clarify liquids, such as oils, tinctures, essences; particularly suitable for use as flat disc filters in Büchner funnels.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge