Gewindeoberteil Service temperature range of PFA is -328F to 500F (-200C to 260C) PFA non-breakable Threaded top for long term storage Fits most standard centrifuge rotors RCF up to 5,000 g 1 * 1 ST
Lieferant: SAVILLEX CORPORATION
Gewindeoberteil Service temperature range of PFA is -328F to 500F (-200C to 260C) PFA non-breakable Threaded top for long term storage Fits most standard centrifuge rotors RCF up to 5,000 g 1 * 1 ST
PRODUKT
Erweitert / Kompakt
Beschreibung Katalognummer Verpackungseinheit Menge
Gewindeoberteil •Service temperature range of PFA is -328°F to 500°F (-200°C to 260°C) •PFA non-breakable •Threaded top for long term storage • Fits most standard centrifuge rotors • RCF up to 5,000 g 1 * 1 ST SAVI210-050-30 1 * 1 ST
Hersteller-Artikelnummer:: 210-050-30
Lokale Artikelnummer:: SAVI210-050-30
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder