Couch cover rolls, Tork
Lieferant: Essity
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
125146 125149 125250 124259
SCAA125146EA 102 EUR InStock
SCAA125146 SCAA125149 SCAA125250 115-3115
Couch cover rolls, Tork
Oberflächenschutz Patientenunterlagen
Hygienic couch cover rolls for protection of patient couches made from strong fibres.
  • Perforated to reduce consumption
  • Tork Universal made from recycled fibres
  • Tork Advanced made from virgin fibres
For use with Tork couch roll dispensers.
Bestellinformation: Available in different materials, one or two ply in multiple lengths and widths.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge