Test tubes without rim, soda lime glass
Lieferant: DWK Life Sciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
A402507000711 A415016000711 A405511000811 A405006500711 A404010500911 A407512000611 A407510000611 A406012000611 A406512000611 A408512000711 A412012000711 A416018001011 A412516000711 A405009500811 A403506500711 A404009500711 A406514500811 A410018001011 A416017001011 A415015750911 A401705000711
SCERA402507000711EA 494 EUR InStock
SCERA402507000711 SCERA415016000711 SCERA405511000811 SCERA405006500711 SCERA404010500911 SCERA407512000611 SCERA407510000611 SCERA406012000611 SCERA406512000611 SCERA408512000711 SCERA412012000711 SCERA416018001011 SCERA412516000711 SCERA405009500811 SCERA403506500711 SCERA404009500711 SCERA406514500811 SCERA410018001011 SCERA416017001011 SCERA415015750911 SCERA401705000711
Test tubes without rim, soda lime glass
Röhrchen Reagenzgläser- und Probenröhrchen
Use the soda lime glass culture tube to view cell cultures in the laboratory.
  • Rimmed top
  • Unlableled
  • Clear soda lime glass
  • Uniform bottom
  • Disposable
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge