Test tubes
Lieferant: DWK Life Sciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
A620025001221 A61001200101 A61601600121 A61801800121 A62003000121 A61001200102 A61601600122 A610016001211
SCERA620025001221EA 4090 EUR InStock
SCERA620025001221 SCERA61001200101 SCERA61601600121 SCERA61801800121 SCERA62003000121 SCERA61001200102 SCERA61601600122 SCERA610016001211
Test tubes
Röhrchen Reagenzgläser- und Probenröhrchen
Borosilicate glass 3,3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
For a wide variety of applications.
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge