SCHRAUBKAPPE SCHWARZ,GEWINDE 15-415, the cap is made phenolic. The liner in the cap is made of Butyl/PTFE. One side is Butyl (red) and the other side is PTFE (grey) 1 * 100 ST
Lieferant: SCHERF PRAZISION
SCHRAUBKAPPE SCHWARZ,GEWINDE 15-415, the cap is made phenolic. The liner in the cap is made of Butyl/PTFE. One side is Butyl (red) and the other side is PTFE (grey) 1 * 100 ST
PRODUKT
Erweitert / Kompakt
Beschreibung Katalognummer Verpackungseinheit Menge
SCHRAUBKAPPE SCHWARZ,GEWINDE 15-415, the cap is made phenolic. The liner in the cap is made of Butyl/PTFE. One side is Butyl (red) and the other side is PTFE (grey) 1 * 100 ST SCERKSSR15415BY100 1 * 100 ST
Hersteller-Artikelnummer:: KSSR15415BY100
Lokale Artikelnummer:: SCERKSSR15415BY100
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder