Microfiltration assembly, ULTRA-WARE®, KIMBLE®
Lieferant: DWK Life Sciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SP179950-4532
SCERSP179950-4532EA 366 EUR InStock
SCERSP179950-4532
Microfiltration assembly, ULTRA-WARE®, KIMBLE®
Filter Membranfiltergeräte
This microfiltration adapter assembly is designed to allow connection of a 47 or 90 mm filtration support base with a straight drip tip to a bottle or reservoir with a GL 45 thread.
Filtrate is collected directly into a media-type bottle when using a standard membrane filtration assembly.
Zertifizierungen: Glass conforms to USP Type I and ASTM E438, Type I, Class A requirements
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge