Centrifuge tubes, oil, graduated, KIMAX®
Lieferant: DWK Life Sciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SP45240-100
SCERSP45240-100EA 725 EUR InStock
SCERSP45240-100
Centrifuge tubes, oil, graduated, KIMAX®
Röhrchen Zentrifugenröhrchen
Borosilicate glass KIMAX®, graduated, snap cap finish.
  • Scale and legend of durable white ceramic enamel
  • Top is tooled to accept a snap cap
It is used in the determination of water and sediment in crude mineral oils, fuel oils and other petroleum products (D1796 and MPMS 10.4 standards), volume of precipitate formed by centrifuging definite quantities of steam cylinder stocks and black oils and other lubricating oils (ASTM D91 and D128), testing for acidity of distillation residues or hydrocarbon liquids of gasoline or petroleum solvents (ASTM D1093).
Tubes are graduated upward to 0,5 ml in divisions of 0,05 ml; from 0,5 ml to 2 ml in divisions of 0,1 ml; from 2 ml to 3 ml in divisions of 0,2 ml; from 3 ml to 5 ml in divisions of 0,5 ml; from 5 ml to 10 ml in divisions of 1 ml; from 10 ml to 25 ml in divisions of 5 ml and at 25 ml to 100 ml in divisions of 25 ml.
Zertifizierungen: Glass conforms to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge